Malachitgrün-Oxalat
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
CAS No.: 2437-29-8
Molecular Formula: C52H54N4O12
Formula Weight: 927.02
Physical Form: Crystalline powder
Appearance: Green
Melting Point: 164° dec.
Beilstein Reference: 3644550
Merck Reference: 14,5699
UN No.: 2811
MDL No.: MFCD00011766
Biological stain, dye reagent, acid base indicator: pH 0.0 yellow, 2.0 green;11.6 green, 14 Colourless
Formel:
C₅₂H₅₄N₄O₁₂ Schmelzpunkt: 164 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011766 CAS-Nummer: 2437-29-8 EINECS: 219-441-7 UN: 2811 ADR: 6.1,III Merck Index: 14,5699 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...