4'-Nitrobenzo-15-krone-5 (4'-Nitrobenzo-1,4,7,10,13-pentaoxacyclopentadecan) ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4-Nitrobenzo-15-crown-5
A16225.06
A16225.06EA
182
EUR
A16225.06
4'-Nitrobenzo-15-krone-5 (4'-Nitrobenzo-1,4,7,10,13-pentaoxacyclopentadecan) ≥99%
4'-Nitrobenzo-15-krone-5 (4'-Nitrobenzo-1,4,7,10,13-pentaoxacyclopentadecan)
Purity: 99%
CAS No.: 60835-69-0
Molecular Formula: C14H19NO7
Formula Weight: 313.30
Melting Point: 93-95°
MDL No.: MFCD00060713
Formel:
C₁₄H₁₉NO₇ Molecular Weight: 313.30 g/mol Schmelzpunkt: 93…95 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00060713 CAS-Nummer: 60835-69-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...