2,4-Dimethoxybenzoesäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

A16252.14 A16252.22
A16252.14EA 37 EUR
A16252.14 A16252.22
2,4-Dimethoxybenzoesäure ≥98%
2,4-Dimethoxybenzoic acid

Purity: 98% CAS No.: 91-52-1 Molecular Formula: C9H10O4 Formula Weight: 182.18 Odour: Odourless Melting Point: 107-111° Beilstein Reference: 2210860 MDL No.: MFCD00002434

Formel: C₉H₁₀O₄
Siedepunkt: 313 °C (1013 hPa)
Schmelzpunkt: 107…111 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002434
CAS-Nummer: 91-52-1
EINECS: 202-074-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR