1,10-Phenanthrolin Hydrochlorid Monohydrat ≥99%

Lieferant: Thermo Fisher Scientific
Gefahr

A16254.06 A16254.14 A16254.22
A16254.06EA 30 EUR
A16254.06 A16254.14 A16254.22
1,10-Phenanthrolin Hydrochlorid Monohydrat ≥99%
1,10-Phenanthrolin Hydrochlorid Monohydrat

Purity: 99% CAS No.: 18851-33-7 Molecular Formula: C12H8N2.HCl.H2O Formula Weight: 234.68 (216.66anhy) Odour: Odourless Melting Point: ca 225° dec. Beilstein Reference: 4007967 UN No.: 2811 MDL No.: MFCD00150061

Formel: C₁₂H₉ClN₂·1H₂O
Dichte: 1,385 g/cm³ (–100.15 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00150061
CAS-Nummer: 18851-33-7
EINECS: 223-325-1
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR