3,4-Dichlorphenylisocyanat ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

A16298.06 A16298.14 AB115816.0005
A16298.06EA 43.3 EUR
A16298.06 A16298.14 ABCRAB115816.0005
3,4-Dichlorphenylisocyanat ≥97%
3,4-Dichlorphenylisocyanat

Purity: 97% CAS No.: 102-36-3 Molecular Formula: C7H3Cl2NO Formula Weight: 188.01 Flash Point: 123°(253°F) Odour: Irritating Boiling Point: 118-120°/18mm Beilstein Reference: 608325 UN No.: 2250 MDL No.: MFCD00002017

Formel: C₇H₃Cl₂NO
Molecular Weight: 188.01 g/mol
Siedepunkt: 118…120 °C (18 mmHg)
Schmelzpunkt: 40…44 °C
Flammpunkt: 123 °C (253 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00002017
CAS-Nummer: 102-36-3
EINECS: 203-026-2
UN: 2250
ADR: 6.1 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR