Pivaloylacetonitrile 99%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 4,4-Dimethyl-3-oxopentanenitrile, 4,4-Dimethyl-3-oxovaleronitrile, Trimethylacetylacetonitrile

A16309.14
A16309.14EA 62.7 EUR
A16309.14
Pivaloylacetonitrile 99%
Pivaloylacetonitrile

Purity: 99% CAS No.: 59997-51-2 Molecular Formula: C7H11NO Formula Weight: 125.17 Melting Point: 66-70° Beilstein Reference: 1746674 UN No.: UN3439 MDL No.: MFCD00010208

Formel: C₇H₁₁NO
Molecular Weight: 125.17 g/mol
Schmelzpunkt: 66…70 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00010208
CAS-Nummer: 59997-51-2
EINECS: 262-017-1
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR