3-Methyl-2-benzothiazolinonehydrazone hydrochloride hydrate 97%

Lieferant: Thermo Fisher Scientific
Gefahr

A16317.06 A16317.14 A16317.22
A16317.06EA 45.9 EUR
A16317.06 A16317.14 A16317.22
3-Methyl-2-benzothiazolinonehydrazone hydrochloride hydrate 97%
3-Methyl-2-benzothiazolinonehydrazone hydrochloride hydrate

Purity: 97% CAS No.: 149022-15-1 Molecular Formula: C8H9N3S.HCl.xH2O Formula Weight: 215.71(anhy) Melting Point: ca 275° dec. Beilstein Reference: 3571041 UN No.: UN2811 MDL No.: MFCD00149370

Formel: C₈H₉N₃S
Schmelzpunkt: ∼ 275 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149370
CAS-Nummer: 149022-15-1
EINECS: 238-428-7
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR