Kaliumacetat ≥99%
Lieferant: Thermo Fisher Scientific
Achtung
|
A16321.0C
A16321.0E
A16321.36
A16321.0CEA
337
EUR
A16321.0C
A16321.0E
A16321.36
Kaliumacetat ≥99%
Kaliumacetat
Purity: 99%
CAS No.: 127-08-2
Molecular Formula: CH3CO2K
Formula Weight: 98.15
Melting Point: 292°
Beilstein Reference: 3595449
Merck Reference: 14,7605
MDL No.: MFCD00012458
Formel:
CH₃COOK Schmelzpunkt: 292 °C Dichte: 1,57 Flammpunkt: >250 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012458 CAS-Nummer: 127-08-2 EINECS: 204-822-2 Merck Index: 14,7605 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...