Quecksilber(II)sulfat ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

A16330.0B A16330.18 A16330.30
A16330.0BEA 596 EUR
A16330.0B A16330.18 A16330.30
Quecksilber(II)sulfat ≥98%

Purity: 98+% CAS No.: 7783-35-9 Molecular Formula: HgO4S Formula Weight: 296.65 Melting Point: dec. Merck Reference: 14,5887 MDL No.: MFCD00011047

Formel: HgSO₄
Dichte: 6,470
Lagertemperatur: Raumtemperatur
MDL: MFCD00011047
CAS-Nummer: 7783-35-9
EINECS: 231-992-5
UN: 1645
ADR: 6.1,II
Merck Index: 14,5887

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR