Cobalt(II)chlorid Hexahydrat ≥98%
Lieferant: Thermo Fisher Scientific
A16346.0B
A16346.18
A16346.30
A16346.0BEA
303
EUR
A16346.0B
A16346.18
A16346.30
Cobalt(II)chlorid Hexahydrat ≥98%
Purity: 98%
CAS No.: 7791-13-1
Molecular Formula: Cl2Co.6H2O
Formula Weight: 237.93 (129.84anhy)
Odour: Odourless
pH: 4.9
Melting Point: 87°
Merck Reference: 14,2437
UN No.: 3077
MDL No.: MFCD00149652
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...