Cobalt(II)chlorid Hexahydrat ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
A16346.0B
A16346.18
A16346.30
A16346.0BEA
303
EUR
A16346.0B
A16346.18
A16346.30
Cobalt(II)chlorid Hexahydrat ≥98%
Cobalt(II)chlorid Hexahydrat
Purity: 98%
CAS No.: 7791-13-1
Molecular Formula: Cl2Co.6H2O
Formula Weight: 237.93 (129.84anhy)
Odour: Odourless
pH: 4.9
Melting Point: 87°
Merck Reference: 14,2437
UN No.: 3077
MDL No.: MFCD00149652
Formel:
CoCl₂·6H₂O Siedepunkt: 110 °C (1013 hPa) Schmelzpunkt: 87 °C Dichte: 1,924 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149652 CAS-Nummer: 7791-13-1 EINECS: 231-589-4 UN: 3077 ADR: 9,III Merck Index: 14,2437 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...