L(+)-Valin
Lieferant: PanReac AppliChem
Synonyme:
(S)-(+)-Valin, (S)-(+)-2-Amino-3-methylbutansäure, (S)-(+)-2-Aminoisovaleriansäure, H-Val-OH
A1637.1000
A1637.1000EA
278
EUR
A1637.1000
L(+)-Valin
L(+)-Valin
Formel:
C₅H₁₁NO₂ Molecular Weight: 117,15 g/mol Siedepunkt: 217 °C (1013 hPa) Schmelzpunkt: 86 °C Dichte: 1,284 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064220 CAS-Nummer: 72-18-4 EINECS: 200-773-6 Merck Index: 13,09458 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...