L(-)-Tryptophan
Lieferant: PanReac AppliChem
A1645.0500
A1645.1000
A1645.0500EA
836
EUR
A1645.0500
A1645.1000
L(-)-Tryptophan
L(-)-Tryptophan
Formel:
C₁₁H₁₂N₂O₂ Schmelzpunkt: 290 °C Dichte: 1,34 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064340 CAS-Nummer: 73-22-3 EINECS: 200-795-6 Merck Index: 12,09929 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...