L(-)-Tryptophan

Lieferant: PanReac AppliChem

Synonyme: L-Tryptophan, Tryptophan, L-α-Amino-3-indolepropionic acid, (S)-2-Amino-3-(3-indolyl)propionic acid, H-Trp-OH

A1645.0500 A1645.1000
A1645.0500EA 836 EUR
A1645.0500 A1645.1000
L(-)-Tryptophan
L(-)-Tryptophan
Formel: C₁₁H₁₂N₂O₂
Molecular Weight: 204.23 g/mol
Schmelzpunkt: 290
Dichte: 1.34 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00064340
CAS-Nummer: 73-22-3
EINECS: 200-795-6
Merck Index: 12,09929

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR