N,N-Dimethyl-1,1,1-trimethylsilylamin 95%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 95%
CAS No.: 2083-91-2
Molecular Formula: C5H15SiN
Formula Weight: 117.27
Flash Point: -19°(-3°F)
Refractive Index: 1.3970
Odour: Amine-like
Boiling Point: 86-87°
Beilstein Reference: 1731861
UN No.: 2733
MDL No.: MFCD00008297
Formel:
C₅H₁₅NSi Siedepunkt: 86…87 °C Dichte: 0,732 Flammpunkt: –19 °C (–3 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008297 CAS-Nummer: 2083-91-2 EINECS: 218-222-3 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...