1-Brom-3-(trifluormethoxy)benzol ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-(Trifluoromethoxy)bromobenzene

A16561.14
A16561.14EA 131 EUR
A16561.14
1-Brom-3-(trifluormethoxy)benzol ≥98%
1-Brom-3-(trifluormethoxy)benzol

Purity: 98+% CAS No.: 2252-44-0 Molecular Formula: C7H4BrF3O Formula Weight: 241.01 Flash Point: 62°(143°F) Refractive Index: 1.4620 Boiling Point: 156-158° Beilstein Reference: 1945938 MDL No.: MFCD00040943

Formel: C₇H₄BrF₃O
Molecular Weight: 241,01 g/mol
Siedepunkt: 156…158 °C
Dichte: 1,620
Flammpunkt: 62 °C (143 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00040943
CAS-Nummer: 2252-44-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR