(3,4-Difluorphenyl)essigsäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3,4-Difluorophenylacetic acid

A16584.06
A16584.06EA 129 EUR
A16584.06
(3,4-Difluorphenyl)essigsäure ≥98%
(3,4-Difluorphenyl)essigsäure

Purity: 98% CAS No.: 658-93-5 Molecular Formula: C8H6F2O2 Formula Weight: 172.13 Flash Point: 110°(230°F) Melting Point: 48-50° Beilstein Reference: 2577768 MDL No.: MFCD00010002

Formel: (C₆H₃F₂)CH₂COOH
Molecular Weight: 172.13 g/mol
Siedepunkt: 259
Schmelzpunkt: 48…50 °C
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00010002
CAS-Nummer: 658-93-5
EINECS: 211-527-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR