Siliciumcarbid, feines Pulver 320 grit
Lieferant: Thermo Scientific
|
Achtung
|
A16601.36
A16601.0E
A16601.0C
A16601.36EA
48.2
EUR
A16601.36
A16601.0E
A16601.0C
Siliciumcarbid, feines Pulver 320 grit
Siliciumcarbid
CAS No.: 409-21-2
Molecular Formula: SiC
Formula Weight: 40.10
Physical Form: Powder
Density: 3.16
Odour: Odourless
MDL No.: MFCD00049531
Formel:
SiC Molecular Weight: 40,1 g/mol Schmelzpunkt: 2700 °C Dichte: 3,16 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00049531 CAS-Nummer: 409-21-2 EINECS: 206-991-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...