Kaliumcarbonat, wasserfrei 99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Pottasche
A16625.0C
A16625.0E
A16625.36
A16625.0CEA
220
EUR
A16625.0C
A16625.0E
A16625.36
Kaliumcarbonat, wasserfrei 99%
Kaliumcarbonat
Purity: 99%
CAS No.: 584-08-7
Molecular Formula: CK2O3
Formula Weight: 138.21
Odour: Odourless
pH: 11,5-12,5
Melting Point: 891°
Beilstein Reference: 4267587
Merck Reference: 14,7619
MDL No.: MFCD00011382
Formel:
K₂CO₃ Schmelzpunkt: 891 °C Dichte: 2,29 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011382 CAS-Nummer: 584-08-7 EINECS: 209-529-3 Merck Index: 14,7619 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...