Lanthan(III)fluorid ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Lanthanum trifluoride

A16673.14 A16673.22
A16673.14EA 53.1 EUR
A16673.14 A16673.22
Lanthan(III)fluorid ≥99%
Lanthan(III)fluorid

Purity: 99% CAS No.: 13709-38-1 Molecular Formula: F3La Formula Weight: 195.90 Odour: Odourless Melting Point: 1493° MDL No.: MFCD00011073

Formel: F₃La
Molecular Weight: 195.9 g/mol
Siedepunkt: ∼2327 °C (1013 hPa)
Schmelzpunkt: 1493 °C
Dichte: 5.936
Lagertemperatur: Raumtemperatur
MDL: MFCD00011073
CAS-Nummer: 13709-38-1
EINECS: 237-252-8
UN: 3288
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR