Bismarckbraun G

Lieferant: Thermo Fisher Scientific

A16674.14 A16674.22
A16674.14EA 35.4 EUR
A16674.14 A16674.22
Bismarckbraun G
Bismarckbraun G

CAS No.: 10114-58-6 Molecular Formula: C18H18N8.2HCl Formula Weight: 419.32 Odour: Odourless Solubility: Soluble in water Merck Reference: 14,1253 MDL No.: MFCD00012977


Plasma stain for mucins, cartilage and goblet cells

Formel: C₁₈H₁₈N8·2HCl
Lagertemperatur: Raumtemperatur
MDL: MFCD00012977
CAS-Nummer: 10114-58-6
EINECS: 233-314-3
Merck Index: 14,1253

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR