Methylrot

Lieferant: Thermo Fisher Scientific

Synonyme: 2-(4-(Dimethylamino)phenylazo)benzoesäure

A16690.14 A16690.22 A16690.36
A16690.14EA 26.8 EUR
A16690.14 A16690.22 A16690.36
Methylrot
Methylrot

CAS No.: 493-52-7 Molecular Formula: C15H15N3O2 Formula Weight: 269.30 Melting Point: 179-182° Beilstein Reference: 750102 Merck Reference: 14,6119 MDL No.: MFCD00002425

Formel: C₁₅H₁₅N₃O₂
Schmelzpunkt: 179…182 °C
Dichte: 1,31 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00002425
CAS-Nummer: 493-52-7
EINECS: 207-776-1
Merck Index: 14,6119

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR