Methylrot
Lieferant: Thermo Fisher Scientific
Synonyme:
2-(4-(Dimethylamino)phenylazo)benzoesäure
A16690.14
A16690.22
A16690.36
A16690.14EA
26.8
EUR
A16690.14
A16690.22
A16690.36
Methylrot
Methylrot
CAS No.: 493-52-7
Molecular Formula: C15H15N3O2
Formula Weight: 269.30
Melting Point: 179-182°
Beilstein Reference: 750102
Merck Reference: 14,6119
MDL No.: MFCD00002425
Formel:
C₁₅H₁₅N₃O₂ Schmelzpunkt: 179…182 °C Dichte: 1,31 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002425 CAS-Nummer: 493-52-7 EINECS: 207-776-1 Merck Index: 14,6119 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...