Alizarin-Komplexon, (max. 10% H₂O)

Lieferant: Thermo Fisher Scientific

A16699.03 A16699.06
A16699.03EA 92.3 EUR
A16699.03 A16699.06
Alizarin-Komplexon, (max. 10% H₂O)
Alizarin-Komplexon

CAS No.: 3952-78-1 Molecular Formula: C19H15NO8 Formula Weight: 385.33 Melting Point: 180-182° Beilstein Reference: 2190028 MDL No.: MFCD00001202

Formel: C₁₉H₁₅NO₈
Schmelzpunkt: 180…182 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00001202
CAS-Nummer: 3952-78-1
EINECS: 223-544-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR