Acrylamid-Bisacrylamid (37.5:1) 30% aqueous solution, crystallized_4

Lieferant: PanReac AppliChem
Gefahr

Synonyme: Acrylamid-Bis

A1672.1000
A1672.1000EA 104 EUR
A1672.1000
Acrylamid-Bisacrylamid (37.5:1) 30% aqueous solution, crystallized_4
Acrylamid-Bis (32:1)
Dichte: 1.03 g/cm³ (20 °C)
MDL: MFCD00080848
UN: 3426
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR