3-(Trifluormethyl)benzylalkohol ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A16739.06
A16739.14
A16739.06EA
42.9
EUR
A16739.06
A16739.14
3-(Trifluormethyl)benzylalkohol ≥97%
3-(Trifluormethyl)benzylalkohol
Purity: 97%
CAS No.: 349-75-7
Molecular Formula: C8H7F3O
Formula Weight: 176.14
Flash Point: 84°(183°F)
Refractive Index: 1.4600
Boiling Point: 79-80°/3mm
Beilstein Reference: 1450161
MDL No.: MFCD00004645
Formel:
C₈H₇F₃O Siedepunkt: 79…80 °C (3 mmHg) Dichte: 1,293 Flammpunkt: 84 °C (183 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004645 CAS-Nummer: 349-75-7 EINECS: 206-489-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...