Benzyltrimethylammoniumchlorid ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

A16759.22 A16759.36
A16759.22EA 25.9 EUR
A16759.22 A16759.36
Benzyltrimethylammoniumchlorid ≥97%
Benzyltrimethylammoniumchlorid

Purity: 97% CAS No.: 56-93-9 Molecular Formula: C6H5CH2N(CH3)3Cl Formula Weight: 185.70 Refractive Index: 1.4440 Odour: Odourless pH: 6-8 Melting Point: ca 238° dec. Beilstein Reference: 3917255 MDL No.: MFCD00011782

Formel: C₁₀H₁₆ClN
Schmelzpunkt: ∼ 238 °C (Zers.)
Dichte: 1,07 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011782
CAS-Nummer: 56-93-9
EINECS: 200-300-3
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR