α-Chlor-3-nitrotoluol ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: 3-Nitrobenzylchlorid, m-Nitrobenzylchlorid, 1-(Chlormethyl)-3-nitrobenzol, α-Chlor-m-nitrotoluol

A16775.09 A16775.18 A16775.30
A16775.09EA 56.5 EUR
A16775.09 A16775.18 A16775.30
α-Chlor-3-nitrotoluol ≥98%
α-Chlor-3-nitrotoluol

Purity: 98% CAS No.: 619-23-8 Molecular Formula: C7H6ClNO2 Formula Weight: 171.58 Flash Point: 110°(230°F) Melting Point: 43-47° Boiling Point: 150°/3mm Beilstein Reference: 742794 UN No.: 3261 MDL No.: MFCD00007272

Formel: C₇H₆ClNO₂
MDL: MFCD00007272
CAS-Nummer: 619-23-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR