Guaifenesin ≥99%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Guajacolglycerinether, 3-(2-Methoxyphenoxy)-1,2-propandiol, 3-(2-Methoxyphenoxy)propan-1,2-diol, 1-(2,3-Dihydroxypropoxy)-2-methoxybenzol
A16827.0B
A16827.0BEA
311
EUR
A16827.0B
Guaifenesin ≥99%
Guaifenesin
Purity: 99+%
CAS No.: 93-14-1
Molecular Formula: C10H14O4
Formula Weight: 198.22
Melting Point: 80-82°
Boiling Point: 215°/19mm
Merck Reference: 14,4555
MDL No.: MFCD00016873
Formel:
C₁₀H₁₄O₄ Molecular Weight: 198,22 g/mol Siedepunkt: 215 °C (19 mmHg) Schmelzpunkt: 80…82 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016873 CAS-Nummer: 93-14-1 EINECS: 202-222-5 Merck Index: 14,4555 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...