4-(Benzyloxy)anilin Hydrochlorid ≥98%
Lieferant: Thermo Fisher Scientific
A16856.14
A16856.22
A16856.14EA
38
EUR
A16856.14
A16856.22
4-(Benzyloxy)anilin Hydrochlorid ≥98%
4-(Benzyloxy)anilin Hydrochlorid
Purity: 98%
CAS No.: 51388-20-6
Molecular Formula: C13H13NO.HCl
Formula Weight: 235.72
Melting Point: ca 225° dec.
Beilstein Reference: 3633307
Formel:
C₁₃H₁₄ClNO Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012995 CAS-Nummer: 51388-20-6 EINECS: 257-170-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...