4-(Benzyloxy)anilin Hydrochlorid ≥98%

Lieferant: Thermo Fisher Scientific

A16856.14 A16856.22
A16856.14EA 38 EUR
A16856.14 A16856.22
4-(Benzyloxy)anilin Hydrochlorid ≥98%
4-(Benzyloxy)anilin Hydrochlorid

Purity: 98% CAS No.: 51388-20-6 Molecular Formula: C13H13NO.HCl Formula Weight: 235.72 Melting Point: ca 225° dec. Beilstein Reference: 3633307

Formel: C₁₃H₁₄ClNO
Lagertemperatur: Raumtemperatur
MDL: MFCD00012995
CAS-Nummer: 51388-20-6
EINECS: 257-170-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR