cis-1,2-Bis(diphenylphosphino)ethylene 97%

Lieferant: Thermo Fisher Scientific
Achtung

A16859.03 A16859.06
A16859.03EA 83.9 EUR
A16859.03 A16859.06
cis-1,2-Bis(diphenylphosphino)ethylene 97%
cis-1,2-Bis(diphenylphosphino)ethylene

Purity: 97% CAS No.: 983-80-2 Molecular Formula: C26H22P2 Formula Weight: 396.41 Melting Point: 122-126° Beilstein Reference: 757792 MDL No.: MFCD00003046

Formel: C₂₆H₂₂P₂
Schmelzpunkt: 122…126 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003046
CAS-Nummer: 983-80-2
EINECS: 213-569-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR