2,6-Di-tert-butyl-p-kresol ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
DBPC, Butylhydroxytoluol, 2,6-Di-tert-butyl-4-methylphenol, Ionol, BHT
A16863.0B
A16863.30
A16863.0BEA
64.6
EUR
A16863.0B
A16863.30
2,6-Di-tert-butyl-p-kresol ≥99%
2,6-Di-tert-butyl-p-kresol
Purity: 99%
CAS No.: 128-37-0
Molecular Formula: C15H24O
Formula Weight: 220.36
Flash Point: 127°(260°F)
Odour: Mild
Boiling Point: 264-265°
Beilstein Reference: 1911640
Merck Reference: 14,1548
UN No.: 3077
MDL No.: MFCD00011644
Formel:
C₁₅H₂₄O Siedepunkt: 264…265 °C Schmelzpunkt: 69…72 °C Dichte: 1,048 Flammpunkt: 127 °C (260 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011644 CAS-Nummer: 128-37-0 EINECS: 204-881-4 ADR: 9,III Merck Index: 14,1548 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...