3-Methylcrotonsäure ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
3,3-Dimethylacrylic acid, 3-Methylbut-2-enoic acid
A16865.14
A16865.14EA
25.7
EUR
A16865.14
3-Methylcrotonsäure ≥98%
3-Methylcrotonsäure
Purity: 98%
CAS No.: 541-47-9
Molecular Formula: C5H8O2
Formula Weight: 100.12
Flash Point: 93°(199°F)
Melting Point: 66-68°
Boiling Point: 198-200°
Beilstein Reference: 1720305
UN No.: 3261
MDL No.: MFCD00004366
Formel:
C₅H₈O₂ Molecular Weight: 100,12 g/mol Siedepunkt: 198…200 °C Schmelzpunkt: 66…68 °C Dichte: 0,969 Flammpunkt: 93 °C (199 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004366 CAS-Nummer: 541-47-9 EINECS: 208-782-7 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...