3,5-Di-tert-butyl-4-hydroxybenzoesäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

A16880.09 A16880.18
A16880.09EA 33.2 EUR
A16880.09 A16880.18
3,5-Di-tert-butyl-4-hydroxybenzoesäure ≥98%
3,5-Di-tert-butyl-4-hydroxybenzoesäure

Purity: 98% CAS No.: 1421-49-4 Molecular Formula: C15H22O3 Formula Weight: 250.34 Melting Point: 206-209° Beilstein Reference: 2216948

Formel: C₁₅H₂₂O₃
Siedepunkt: 374 °C (1013 hPa)
Schmelzpunkt: 206…209 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00008827
CAS-Nummer: 1421-49-4
EINECS: 215-823-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR