Bromphenolblau Natriumsalz

Lieferant: Thermo Fisher Scientific

A16899.09 A16899.18
A16899.09EA 32.5 EUR
A16899.09 A16899.18
Bromphenolblau Natriumsalz
Bromphenolblau Natriumsalz

CAS No.: 34725-61-6 Molecular Formula: C19H9Br4NaO5S Formula Weight: 691.96 Melting Point: 300° Merck Reference: 14,1444 MDL No.: MFCD00013793

Formel: C₁₉H₉Br₄NaO₅S
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00013793
CAS-Nummer: 34725-61-6
EINECS: 252-170-2
Merck Index: 14,1444

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR