(R)-(+)-1-(1-Naphthyl)ethylamin 99%

Lieferant: Thermo Fisher Scientific
Gefahr

A16918.03 A16918.06 A16918.14
A16918.03EA 40.8 EUR
A16918.03 A16918.06 A16918.14
(R)-(+)-1-(1-Naphthyl)ethylamin 99%
(R)-(+)-1-(1-Naphthyl)ethylamin

Purity: 99% CAS No.: 3886-70-2 Molecular Formula: C12H13N Formula Weight: 171.24 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.6230 Odour: Amine-like Boiling Point: 152-153°/11mm Beilstein Reference: 2208025 UN No.: 2922 MDL No.: MFCD00064114

Formel: C₁₂H₁₃N
Siedepunkt: 152…153 °C (11 mmHg)
Dichte: 1,060
Flammpunkt: >110 °C (230 °F)
MDL: MFCD00064114
CAS-Nummer: 3886-70-2
EINECS: 223-425-5
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR