Trisodium 2-(4-Sulphophenylazo)-1,8-dihydroxynaphthalene-3,6-disulphonate (SPADNS)

Lieferant: Thermo Scientific
Achtung

Synonyme: SPADNS, 2-(4-Sulphophenylazo)chromotropic acid trisodium salt, 4,5-Dihydroxy-3-(4-sulphophenylazo)-2,7-naphthalenedisulphonic acidtrisodium salt, 1,8-Dihydroxy-2-(4-sulphophenylazo)naphthalene-3,6-disulphonic acidtrisodium salt, Sulphanilic acid azochromotrope, 2-(p-sulphophenylazo)-1,8-Dihydroxy-3,6-Naphthalenedisulphonic acidtrisodium salt, Trisodium 1,8-Dihydroxy-2-(4-sulphophenylazo)naphthalene-3,6-disulphonate

A16933.09
A16933.09EA 29 EUR
A16933.09
Trisodium 2-(4-Sulphophenylazo)-1,8-dihydroxynaphthalene-3,6-disulphonate (SPADNS)
Trisodium 2-(4-Sulphophenylazo)-1,8-dihydroxynaphthalene-3,6-disulphonate (SPADNS)

CAS No.: 23647-14-5 Molecular Formula: C16H9N2Na3O11S3 Formula Weight: 570.41 Melting Point: 300° MDL No.: MFCD00003952

Formel: C₁₆H₁₂N₂O₁₁S₃
Molecular Weight: 570,40 g/mol
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003952
CAS-Nummer: 23647-14-5
EINECS: 245-803-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR