Kaliumhexacyanoferrat(III) ≥98%
Lieferant: Thermo Fisher Scientific
A16946.0B
A16946.0I
A16946.30
A16946.0BEA
82.9
EUR
A16946.0B
A16946.0I
A16946.30
Kaliumhexacyanoferrat(III) ≥98%
Purity: 98+%
CAS No.: 13746-66-2
Molecular Formula: C6FeK3N6
Formula Weight: 329.26
Melting Point: dec.
Merck Reference: 14,7630
MDL No.: MFCD00011392
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...