Adeninsulfat ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Adeninhemisulfat

A16964.09 A16964.18
A16964.09EA 39.9 EUR
A16964.09 A16964.18
Adeninsulfat ≥98%
Adeninsulfat

Purity: 98+% CAS No.: 321-30-2 Molecular Formula: C10H10N10.H2SO4 Formula Weight: 368.34 Odour: Odourless Melting Point: ca 210° dec. Beilstein Reference: 3820263 Merck Reference: 14,152 MDL No.: MFCD00213655

Formel: C₅H₅N₅·½H₂SO₄
Schmelzpunkt: ∼ 210 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00213655
CAS-Nummer: 321-30-2
EINECS: 206-286-5
Merck Index: 14,152

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR