2,3-Difluorphenol ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
A16971.03
A16971.06
A16971.03EA
30.6
EUR
A16971.03
A16971.06
2,3-Difluorphenol ≥98%
2,3-Difluorphenol
Purity: 98+%
CAS No.: 6418-38-8
Molecular Formula: C6H4F2O
Formula Weight: 130.09
Flash Point: 56°(132°F)
Odour: Phenol-like
Boiling Point: 53-55°/25mm
Beilstein Reference: 2082265
UN No.: 2921
MDL No.: MFCD00010262
Formel:
C₆H₄F₂O Siedepunkt: 53…55 °C (25 mmHg) Schmelzpunkt: 34…38 °C Flammpunkt: 56 °C (132 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010262 CAS-Nummer: 6418-38-8 UN: 2921 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...