Lithiumdihydrogenorthophosphat ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

A16987.0B A16987.30
A16987.0BEA 196 EUR
A16987.0B A16987.30
Lithiumdihydrogenorthophosphat ≥97%
Lithium dihydrogen orthophosphate

Purity: 97% CAS No.: 13453-80-0 Molecular Formula: LiH2PO4 Formula Weight: 103.93 Odour: Odourless Melting Point: 100° MDL No.: MFCD00016188

Formel: H₂LiO₄P
Schmelzpunkt: >100 °C
Dichte: 2,461
Lagertemperatur: Raumtemperatur
MDL: MFCD00016188
CAS-Nummer: 13453-80-0
EINECS: 236-633-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR