L-Arginin Hydrochlorid
Lieferant: PanReac AppliChem
A1700.1000
A1700.1000EA
156
EUR
A1700.1000
L-Arginin Hydrochlorid
L-Arginin Hydrochlorid
Formel:
C₆H₁₅ClN₄O₂ Schmelzpunkt: 218 °C Dichte: 1,42 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064550 CAS-Nummer: 1119-34-2 EINECS: 214-275-1 Merck Index: 13,00785 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...