Ammoniumbismutcitrat Bi 48-52%, (max. 2% H₂O)

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Bismuth Ammonium Citrate, Citric acid ammonium bismuth salt

A17003.30 A17003.18 A17003.0B
A17003.30EA 181 EUR
A17003.30 A17003.18 A17003.0B
Ammoniumbismutcitrat Bi 48-52%, (max. 2% H₂O)
Ammoniumbismutcitrat

CAS No.: 25530-63-6 Molecular Formula: C6H10BiNO8 Formula Weight: 433.12 MDL No.: MFCD00036420

Formel: C₆H₁₁BiNO₇
Lagertemperatur: Raumtemperatur
MDL: MFCD00036420
CAS-Nummer: 25530-63-6
EINECS: 247-078-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR