Natriumhydrogencarbonat ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

A17005.0C A17005.0E A17005.36
A17005.0CEA 163 EUR
A17005.0C A17005.0E A17005.36
Natriumhydrogencarbonat ≥99%
Natriumhydrogencarbonat

Purity: 99% CAS No.: 144-55-8 Molecular Formula: NaHCO3 Formula Weight: 84.01 Refractive Index: 1.500 Odour: Odourless pH: 8,1-8,4 Beilstein Reference: 4153970 Merck Reference: 14,8583 MDL No.: MFCD00003528

Formel: NaHCO₃
Schmelzpunkt: 270 °C
Dichte: 2,159
Lagertemperatur: Raumtemperatur
MDL: MFCD00003528
CAS-Nummer: 144-55-8
EINECS: 205-633-8
Merck Index: 14,8583
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6b1b80d2afb3deab0c4ca836a921fcaad637890&vid=1163b37d9cf1c10fe0ed252e02a51fe12d6b50dd&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR