CAPS (3-(Cyclohexylamino)propansulfonsäure) ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3-Cyclohexylaminopropanesulphonic acid
A17037.14
A17037.22
A17037.36
A17037.14EA
34.3
EUR
A17037.14
A17037.22
A17037.36
CAPS (3-(Cyclohexylamino)propansulfonsäure) ≥99%
CAPS (3-(Cyclohexylamino)propansulfonsäure)
Purity: 99%
CAS No.: 1135-40-6
Molecular Formula: C9H19NO3S
Formula Weight: 221.32
Odour: Odourless
Melting Point: 324°
Beilstein Reference: 2835588
Merck Reference: 14,1767
MDL No.: MFCD00003837
Formel:
C₆H₁₁NH(CH₂)₃SO₃H Molecular Weight: 221.32 g/mol Schmelzpunkt: 324 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003837 CAS-Nummer: 1135-40-6 EINECS: 214-492-1 Merck Index: 13,01773 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...