DL-α-Tocopherol ≥97%

Lieferant: Thermo Fisher Scientific

Synonyme: (±)-α-Tocopherol, all-rac-α-Tocopherol, 3,4-Dihydro-2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-2H-benzopyran-6-ol, Vitamin E Alkohol, Vitamin E

A17039.0B A17039.18 A17039.30
A17039.0BEA 526 EUR
A17039.0B A17039.18 A17039.30
DL-α-Tocopherol ≥97%
DL-α-Tocopherol

Purity: 97+% CAS No.: 10191-41-0 Molecular Formula: C29H50O2 Formula Weight: 430.72 Flash Point: 240°(464°F) Physical Form: Viscous Liquid Density: 0.950 Refractive Index: 1.5050 Sensitivity: Light Sensitive Melting Point: 2-4° Boiling Point: 200-220°/0.1mm Beilstein Reference: 94012 Merck Reference: 14,10021 MDL No.: MFCD00072051

Formel: C₂₉H₅₀O₂
Molecular Weight: 430.71 g/mol
Siedepunkt: 200…220 °C (0.1 mmHg)
Schmelzpunkt: 2…4 °C
Dichte: 0.950
Flammpunkt: 240 °C (464 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00072051
CAS-Nummer: 10191-41-0
EINECS: 233-466-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR