DL-α-Tocopherol ≥97%
Lieferant: Thermo Fisher Scientific
Synonyme:
(±)-α-Tocopherol, all-rac-α-Tocopherol, 3,4-Dihydro-2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-2H-benzopyran-6-ol, Vitamin E Alkohol, Vitamin E
A17039.0B
A17039.18
A17039.30
A17039.0BEA
526
EUR
A17039.0B
A17039.18
A17039.30
DL-α-Tocopherol ≥97%
DL-α-Tocopherol
Purity: 97+%
CAS No.: 10191-41-0
Molecular Formula: C29H50O2
Formula Weight: 430.72
Flash Point: 240°(464°F)
Physical Form: Viscous Liquid
Density: 0.950
Refractive Index: 1.5050
Sensitivity: Light Sensitive
Melting Point: 2-4°
Boiling Point: 200-220°/0.1mm
Beilstein Reference: 94012
Merck Reference: 14,10021
MDL No.: MFCD00072051
Formel:
C₂₉H₅₀O₂ Molecular Weight: 430.71 g/mol Siedepunkt: 200…220 °C (0.1 mmHg) Schmelzpunkt: 2…4 °C Dichte: 0.950 Flammpunkt: 240 °C (464 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00072051 CAS-Nummer: 10191-41-0 EINECS: 233-466-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...