L(+)-Lysin Monohydrochlorid

Lieferant: PanReac AppliChem

A1706.1000
A1706.1000EA 98.1 EUR
A1706.1000
L(+)-Lysin Monohydrochlorid
L(+)-Lysin Monohydrochlorid
Formel: H₂N(CH₂)₄CH(NH₂)CO₂H·HCl
Schmelzpunkt: 263…264 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00064564
CAS-Nummer: 657-27-2
EINECS: 211-519-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR