2-Amino-5-(trifluormethyl)-1,3,4-thiadiazol 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Amino-5-trifluoromethyl-1,3,4-thiadiazole
A17061.03
A17061.03EA
26.9
EUR
A17061.03
2-Amino-5-(trifluormethyl)-1,3,4-thiadiazol 98%
2-Amino-5-(trifluoromethyl)-1,3,4-thiadiazole
Purity: 98%
CAS No.: 10444-89-0
Molecular Formula: C3H2F3N3S
Formula Weight: 169.13
Melting Point: 222-227°
Beilstein Reference: 513140
Formel:
C₃H₂F₃N₃S Molecular Weight: 169,13 g/mol Schmelzpunkt: 222…227 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003109 CAS-Nummer: 10444-89-0 EINECS: 233-930-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...